missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD6A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38027-20ul
This item is not returnable.
View return policy
Description
PARD6A Polyclonal antibody specifically detects PARD6A in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PARD6A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| PAR-6, PAR-6 alpha, par-6 partitioning defective 6 homolog alpha (C. elegans), PAR6A, PAR6alpha, PAR-6Apar-6 (partitioning defective 6, C.elegans) homolog alpha, PAR6C, partitioning defective 6 homolog alpha, partitioning defective-6 homolog alpha, partitioning-defective protein 6, Tax interaction protein 40, TAX40, Tax-interacting protein 40, TIP-40PAR6 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PARD6A (Q9NPB6).,, Sequence:, MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKRAEADS | |
| 20 μL | |
| Cell Cycle and Replication | |
| 50855 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction