missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD3B, Mouse anti-Human, Clone: 8D7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00117583-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: SPDAFETEVAAQLAAFKPIGGEIEVTPSALKLGTPLLVRRSSDPVPGPPADTQPSASHPGGQSLKLVVPDSTQNLEDREVLNGVQTELLTSPRTKDTLSDMTRTVEISGSpecifications
| PARD3B | |
| Monoclonal | |
| Unconjugated | |
| par-3 partitioning defective 3 homolog B (C. elegans) | |
| ALS2CR19/MGC16131/PAR3B/PAR3L/PAR3LC/PAR3beta/Par3Lb | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 8D7 | |
| In 1x PBS, pH 7.4 | |
| NM_152526 | |
| PARD3B | |
| ALS2CR19 (NP_689739, 101 a.a. ∼ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 117583 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction