missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD3B Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-97771
This item is not returnable.
View return policy
Description
PARD3B Polyclonal specifically detects PARD3B in Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| PARD3B | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ALS2CR19, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 19, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein, MGC16131, par-3 partitioning defective 3 homolog B (C. elegans), PAR3B, PAR3beta, PAR3-beta, PAR3L, PAR3-L protein, Par3Lb, PAR3LC, partitioning defective 3 homolog B, Partitioning defective 3-like protein, partitioning-defective 3-beta | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_001074519.2 | |
| Pard3b | |
| Peptide with sequence C-RNQDPRQKNP corresponding to C-Terminus according to Uniprot Mouse NP_001074519.2. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 117583 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction