missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARD3/Par3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£196.00 - £558.00
Specifications
| Antigen | PARD3/Par3 |
|---|---|
| Concentration | 0.5 mg/ml |
| Dilution | Western Blot 1:250 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18432781
|
Novus Biologicals
NBP2-11253 |
0.1 mg |
£558.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18038158
|
Novus Biologicals
NBP2-11253-0.025MG |
0.025 mg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PARD3/Par3 Polyclonal specifically detects PARD3/Par3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| PARD3/Par3 | |
| Western Blot 1:250 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FLJ21015, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog | |
| Synthetic peptide from amino acids 31-342 corresponding to rat PAR2 protein from the following sequence, was selected from the unique region of the mouse PAR3 protein, peptide was post-synthetically modified to achieve highest antigenicity before used for coupling to KLH using heterobifunctional cross linker for immunogen preparation. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. | |
| 56288 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title