missing translation for 'onlineSavingsMsg'
Learn More

PARD3/Par3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18432781 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432781 0.1 mg 0.1mg
18038158 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18432781 Supplier Novus Biologicals Supplier No. NBP211253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PARD3/Par3 Polyclonal specifically detects PARD3/Par3 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARD3/Par3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:250
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%.
Gene Alias FLJ21015, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog
Host Species Rabbit
Immunogen Synthetic peptide from amino acids 31-342 corresponding to rat PAR2 protein from the following sequence, was selected from the unique region of the mouse PAR3 protein, peptide was post-synthetically modified to achieve highest antigenicity before used for coupling to KLH using heterobifunctional cross linker for immunogen preparation.
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56288
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.