missing translation for 'onlineSavingsMsg'
Learn More

Parbendazole, TRC

Product Code. 30132832
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132832 1 g 1g
30007099 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132832 Supplier Toronto Research Chemicals Supplier No. TRCP1932601G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14255-87-9
Molecular Formula C13 H17 N3 O2
Molecular Weight (g/mol) 247.29
Synonym Carbamic acid, N-(6-butyl-1H-benzimidazol-2-yl)-, methyl ester, 2-Benzimidazolecarbamic acid, 5-butyl-, methyl ester (8CI), Carbamic acid, (5-butyl-1H-benzimidazol-2-yl)-, methyl ester (9CI), 5-Butyl-2-(carbomethoxyamino)benzimidazole, Helatac, Helmatac, Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate, Methyl 5(6)-butyl-2-benzimidazolecarbamate, Methyl 5-butylbenzimidazole-2-carbamate, PBZ, PBZ (fungicide), Parbendazole, SKF 29044
IUPAC Name methyl N-(6-butyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCCc1ccc2nc(NC(=O)OC)[nH]c2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H17N3O2/c1-3-4-5-9-6-7-10-11(8-9)15-12(14-10)16-13(17)18-2/h6-8H,3-5H2,1-2H3,(H2,14,15,16,17)
Formula Weight 247.1321
Percent Purity >95%
Chemical Name or Material Parbendazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.