missing translation for 'onlineSavingsMsg'
Learn More

Parathyroid Hormone Rabbit, Clone: PTH/1717R, enQuireBio™

Rabbit Monoclonal Antibody

£181.83 - £423.29

Specifications

Antigen Parathyroid Hormone
Clone PTH/1717R
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15900738
View Documents
EnquireBio
5741-RBM7-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15910738
View Documents
EnquireBio
5741-RBM7-P1
100 μg
£423.29
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Parathyroid Hormone
200μg/mL
Monoclonal
Rabbit
10mM PBS with or without 0.05% BSA and 0.05% azide.
5741
Recombinant human PTH protein N-terminal fragment
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
9kDa
PTH/1717R
Immunohistochemistry
Unconjugated
RUO
NP_001303281
PTH
IgG κ
Protein A/G Purified
Cytoplasmic and Secreted
Human parathyroid gland carcinoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.