missing translation for 'onlineSavingsMsg'
Learn More

Parathion 1000 μg/mL in n-Hexane, Dr. Ehrenstorfer

Product Code. 30316240
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316240 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316240 Supplier Dr. Ehrenstorfer Supplier No. DREYS09010031HE
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Parathion-ethyl
CAS 56-38-2
Analyte or Component Names Parathion-ethyl
Shipping Condition Room Temperature
Molecular Formula C10 H14 N O5 P S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphorothioic acid, O,O-diethyl O-(4-nitrophenyl) ester, Phosphorothioic acid, O,O-diethyl O-(p-nitrophenyl) ester (6CI,8CI), AAT, AATP, Alkron, Alleron, American Cyanamid 3422, Aphamite, Aralo, BAY 1605, Bayer E-605, Bladan F, DNTP, Diethyl 4-nitrophenyl phosphorothioate, Diethyl p-nitrophenyl phosphorothionate, Diethyl p-nitrophenyl thionophosphate, Diethyl parathion, E 605, E 605F, E 605FT20, ENT 15108, Ecatox 20, Ekatox, Ekatox 20, Ethyl parathion, Etilon, Folidol, Folidol E, Folidol E-605, Folidol oil, Fosferno, Fostox, Galpar, Gearphos, Lirothion, NIUIF 100, NSC 8933, Niran, Nitrostigmine, Nourithion, O,O-Diethyl O-(4-nitrophenyl) phosphorothioate, O,O-Diethyl O-(p-nitrophenyl) phosphorothioate, O,O-Diethyl O-p-nitrophenyl thiophosphate, Oleofos 20, Oleoparathene, Oleoparathion, Pacol, Paramar 50, Paraphos, Parathene, Parathion, Parathion A, Parathion-ethyl, Penncap E, RB, Rhodiasol, Rhodiatox, Selephos, Super Rodiatox, Thiomex, Thiophos, Thiophos 3422, Vitrex
InChI Formula InChI=1S/C10H14NO5PS/c1-3-14-17(18,15-4-2)16-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3
SMILES CCOP(=S)(OCC)Oc1ccc(cc1)[N+](=O)[O-]
IUPAC Name diethoxy-(4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane
Solvent or Matrix n-Hexane
Industry Type Food Testing
Molecular Weight (g/mol) 291.26
Quantity 5 x 1 mL
Formula Weight 291.033 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.