missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline Hydrochloride, TRC

Product Code. 30132519
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132519 25 g 25g
30138694 50 g 50g
30004244 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132519 Supplier Toronto Research Chemicals Supplier No. TRCP19326825G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19 H17 N3 . Cl H
Molecular Weight (g/mol) 323.82
Synonym Benzenamine, 4,4'-[(4-imino-2,5-cyclohexadien-1-ylidene)methylene]bis-, monohydrochloride (9CI), Benzenamine, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-, monohydrochloride, C.I. Basic Red 9, monohydrochloride (8CI), Para Magenta (6CI), Basic Parafuchsine, Basic Red 9, C.I. 42500, C.I. Basic Red 9, Calcozine Magenta N, Fuchsine DR 001, Fuchsine SP, Fuchsine SPC, NSC 10460, Orient Para Magenta Base, Parafuchsin, Parafuchsine, Pararosaniline, Pararosaniline chloride, Pararosaniline hydrochloride, p-Fuchsin, p-Fuchsine, p-Rosaniline hydrochloride
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES Cl.Nc1ccc(cc1)C(=C2C=CC(=N)C=C2)c3ccc(N)cc3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C19H17N3.ClH/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;/h1-12,20H,21-22H2;1H
Formula Weight 323.1189
Chemical Name or Material Basic Red 9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.