missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pararosaniline chloride, pure, high purity biological stain

Product Code. 11926931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11926931 500 g Glass bottle 500g
10390934 5 g Glass bottle 5g
10104620 25 g Glass bottle 25g
10571671 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11926931 Supplier Thermo Scientific Chemicals Supplier No. 227885000
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00001657
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 268.0°C to 270.0°C
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol
Formula Weight 323.83
Grade Pure
Packaging Glass bottle
Quantity 500 g
Chemical Name or Material Pararosaniline chloride
compliance-icons
Product Identifier
  • Pararosaniline chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.