missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pararosaniline chloride, pure, high purity biological stain

Product Code. 10104620 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10104620

Brand: Thermo Scientific Chemicals 227880250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00001657
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 268.0°C to 270.0°C
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol
Formula Weight 323.83
Grade Pure
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Pararosaniline chloride
compliance-icons
Product Identifier
  • Pararosaniline chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.