missing translation for 'onlineSavingsMsg'
Learn More

Paraquat Dichloride (42% w/w in Water), TRC

Product Code. 30408854
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408854 10 g -
30412555 2.5 g 2.5g
30418762 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408854 Supplier Toronto Research Chemicals Supplier No. TRCP19192010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1910-42-5
Molecular Formula C12 H14 N2 . 2 Cl
Molecular Weight (g/mol) 257.16
Synonym 4,4'-Bipyridinium, 1,1'-dimethyl-, chloride (1:2), 1,1'-Dimethyl-4,4'-bipyridinium dichloride (6CI,7CI), 4,4'-Bipyridinium, 1,1'-dimethyl-, dichloride (8CI,9CI), 1,1'-Dimethyl-4,4'-dipyridinium dichloride, 1,1'-Dimethyl-4,4'-dipyridylium dichloride, AH 501, AH 501 (herbicide), Cekuquat, Cyclone Max, Dimethyl viologen chloride, Galokson, Gramixel, Gramoxone, Gramoxone D, Gramoxone Inteon, Gramoxone Max, Gramoxone S, Gramoxone W, Gramoxone dichloride, Methyl viologen, Methyl viologen dichloride, Methylviologen chloride, N,N'-Dimethyl-4,4'-bipyridinium dichloride, N,N'-Dimethyl-4,4'-dipyridylium dichloride, Nuquat, OK 622, Paragreen A, Paraquat, Paraquat dichloride
IUPAC Name 1-methyl-4-(1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
SMILES [Cl-].[Cl-].C[n+]1ccc(cc1)c2cc[n+](C)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H14N2.2ClH/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;/h3-10H,1-2H3;2*1H/q+2;;/p-2
Formula Weight 256.0534
Concentration or Composition (by Analyte or Components) 42%
Chemical Name or Material Paraquat dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.