missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paramyxovirus SV5 Pk Antibody (SV5-Pk1), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62264MFV500
This item is not returnable.
View return policy
Description
Paramyxovirus SV5 Pk Monoclonal antibody specifically detects Paramyxovirus SV5 Pk in Mouse, Virus samples. It is validated for Flow Cytometry
Specifications
| Paramyxovirus SV5 Pk | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Mouse, Virus | |
| Purified |
| Flow Cytometry | |
| SV5-Pk1 | |
| Flow Cytometry | |
| Simian virus 5; SPV5gp2; V protein | |
| Mice were infected with the paramyxovirus SV5, Simian-Virus 5 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction