missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Paralemmin 3 Polyclonal antibody specifically detects Paralemmin 3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Paralemmin 3 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | PALM3, paralemmin-3 |
| Host Species | Rabbit |
| Immunogen | Paralemmin3 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human Paralemmin3. The immunogen is located within amino acids 50 - 100 of Paralemmin3. Amino Acid Squence: PEPSEDPTSKDPQSPEGQA |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?