missing translation for 'onlineSavingsMsg'
Learn More

Paralemmin 3 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18403008 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403008 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403008 Supplier Novus Biologicals Supplier No. NBP281979G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Paralemmin 3 Polyclonal antibody specifically detects Paralemmin 3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paralemmin 3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias PALM3, paralemmin-3
Host Species Rabbit
Immunogen Paralemmin3 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human Paralemmin3. The immunogen is located within amino acids 50 - 100 of Paralemmin3. Amino Acid Squence: PEPSEDPTSKDPQSPEGQA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 342979
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.