missing translation for 'onlineSavingsMsg'
Learn More

Paraldehyde, 99.5%

Product Code. 15254694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15254694 10 L 10 L
10164730 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254694 Supplier Thermo Scientific Acros Supplier No. 275080100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-63-7 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-63-7
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
InChI Key SQYNKIJPMDEDEG-UHFFFAOYSA-N
IUPAC Name 2,4,6-trimethyl-1,3,5-trioxane
SMILES CC1OC(C)OC(C)O1

Specifications

Melting Point 12.0°C
Density 0.9900 g/mL
Boiling Point 124.0°C
Flash Point 24°C
Assay Percent Range 99.4 % min. (GC)
Refractive Index 1.4035 to 1.4055
Quantity 10 L
Beilstein 19, 385
Merck Index 15, 7131
Specific Gravity 0.99
Solubility Information Solubility aq. soln.: 125 g/L (25°C). Other solubilities: miscible with most common organic solvents
Viscosity 1.31 mPa.s (20°C)
Formula Weight 132.16
Chemical Name or Material Paraldehyde
Show More Show Less
compliance-icons
Product Identifier
  • Paraldehyde
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.