missing translation for 'onlineSavingsMsg'
Learn More

Paraldehyde, 99.5%

Product Code. 10319233 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 10319233

Brand: Thermo Scientific Acros 275080025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paraldehyde, 99.5% 2.5L | CAS: 123-63-7 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Paraldehyde, 99.5% 2.5L Plastic Bottle Cas No. 123-63-7
TRUSTED_SUSTAINABILITY

Specifications

CAS 123-63-7
Melting Point 12°C
Boiling Point 65°C to 82°C
Packaging PLASTIC
Molecular Formula C6H12O3
Quantity 2.5 L
InChI Key SQYNKIJPMDEDEG-UHFFFAOYSA-N
SMILES CC1OC(C)OC(C)O1
IUPAC Name 2,4,6-trimethyl-1,3,5-trioxane
Molecular Weight (g/mol) 132.16
Percent Purity 99.5%
Show More Show Less
compliance-icons
Product Identifier
  • Paraldehyde
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.