missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cold Filter Plugging Point Standard, Paragon Scientific™
Click to view available options
Quantity:
250 mL
Description
- Certified by Round Robin Method by independent test laboratories
- Supplied in tamper evident UN compliant security pack
- Pack size 250mL
- 12 months shelf life
Specifications
Quantity | 250 mL |
Safety and Handling
Product Identifier
- CRM-CFGO
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Aspiration toxicity Category 1
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
Nominal values quoted
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'