missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, 96%, extra pure

Product Code. 10443361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 kg
10 kg
Packaging:
Plastic Bucket
Plastic bottle
Unit Size:
10kg
1kg
25g
3kg
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10443361 3 kg Plastic bottle 3kg
10351641 25 g Plastic bottle 25g
10424131 500 g Plastic bottle 500g
10342243 1 kg Plastic bottle 1kg
11924801 10 kg Plastic Bucket 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10443361 Supplier Thermo Scientific Acros Supplier No. 416780030
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning | CAS: 30525-89-4 | CH2O | 30.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Decomposition Information 260°C
Melting Point 120.0° to 170.0°C
Color White
Density 1.03 (Air = 1.0)
pH 4.0 to 5.5 (10% suspension)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Linear Formula (CH2O)n
Quantity 3 kg
UN Number 2213
Solubility Information Solubility in water: sparingly soluble. Other solubilities: the higher polymers are insoluble in water, insoluble in alcohol and ether
Vapor Pressure 1.2mmHg at 25°C
Percent Purity 96%
Grade Extra Pure
Physical Form Beads or Powder
Chemical Name or Material Paraformaldehyde, Extra Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Paraformaldehyde

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.