missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 20% w/v aq. soln., methanol free

Product Code. 16875201
Change view
Click to view available options
Quantity:
10 x 10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16875201 10 x 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16875201 Supplier Thermo Scientific Chemicals Supplier No. 047340.9M
  • ONLINE EXCLUSIVE PRICE
    £77.60
    £69.65 / Pack of 1
    Save £7.95
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solution is micro-filtered and supplied in ampules. | CAS: 50-00-0 | CH2O | 30.03 g/mol

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Linear Formula (CH2O)n
Quantity 10 x 10 mL
Beilstein 102769
Merck Index 14,7025
Solubility Information Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol.
Concentration or Composition (by Analyte or Components) 20% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Paraformaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.