missing translation for 'onlineSavingsMsg'
Learn More

Paraffin Wax, TRC

Product Code. 30011263
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011263 500 g 500g
30009565 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011263 Supplier Toronto Research Chemicals Supplier No. TRCP191410500G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-74-2
Synonym Paraffin, Paraffin hard, Paraffin light liquid, Paraffin liquid, Paraffin white soft, Paraffin yellow soft, Hydrocarbon waxes, Paraffin wax and other hydrocarbon waxes, Paraffin waxes and Hydrocarbon waxes, 140F, 150F, 2202A, 431B, 45A, 45A (wax), 4656A, Advawax 165, Akrowax 5030, Akrowax 5031, Akrowax 5032, Altafin 125, AmpliWax, Ampliwax Gem 50, Anti-Chek, Apiezon M, Apiezon N, Apiezon W, Apiezon Wax W 100, AquaBead 325E, Aquamol 1200, Arcowax 1150G, Arcowax 2143G, Arcowax 4154G, Arcowax 4158G, Aristowax, Aristowax 125, Aristowax 143, Aristowax 165, Bareco 170/175, Bareco PX 100, Basewax H 187, Boler 1328, Boler 1397, Boler 1421, Boler 1977, Boler 941, Bondwax WE 70, CRW 141, Ceratak, Ceretal 165, Chevron 143, Chevron 159, Chevron 4042, Cosmoloid 80H, Crolene LC, DMW 6064, DMW 7074, DP 652, DW 5456, DW 5658, Diaproof, Durowax FT 300, Dyedit C 30, Dyedit EK, Dyedit S 8, EM 046H, EM 048H, EM 061H, Ecco Wax 244, Eskar R 25, Eskar R 45, Esso 3150, Esso 778, Esso 778E, Evoral PL, Evoral SP, Excerex HW 26502PE, Excerex HW 41420P, Excerex HW 48070B, FT 180, FT 300, FTP 1005, Flexowax C, Freeman 155/160, G-C Base Plate Paraffin and Wax, G-C Base Plate Paraffin and Wax-Normal, GRF, Gatch, Gulfwax, H 1N3, H 1N6, HI, HI-Mic 2090, HM-FRW 156, HNP 1, HNP 1 (wax), HNP 12, HNP 14, HNP 14G, HNP 16, HS 28W, HUB 3-1, Harowax L 1, Harowax L 2, Hidorin D 338, Hidorin E 139, Hidorin L 703-35, Histowax HX 0482-5, Hydrocarbons, waxes, Hytec B 5936, IGI 411, Indramic 30, Irgawax 366, J 1440, Johnsons Wax 111, K 51, K 51 (wax), K 61, K 61 (wax), KW 622, Kerawax 422, Kogasin III, LLN, LM-FRW 126, Lenolene AC, Luvax 1266, Luvax 2191, ME 47950, ML 445, MM-RHC 135, Masterkure 122, Meghwax CPB 500, Mobil 150, Mobil 150-155F, Mobilcer 161, Mobilcer 216, Mobilcer 739, Mobilcer ED 80/229, Mobilwax 140, Mobilwax 145, Mobilwax 220, Mulrex 114, Mystolene MK 7, Mystolene SP 30, N 481-0705-3, N 481-1022-2, N 623, N 624, N 627, N 900, NC 511, NNP 9, NV 120, NV 120 (hydrocarbon), NXK 501S, Neowax LA 05, Nimi Pink Base Plate Paraffin and Wax, Nisseki 145, Noda Wax NC 511, Nopalets MP 80, Nopalets MP 85, Nopalets MP 90, Nopcosize DS 101, Norane FH, OT 78, Okerin 11131, Okerin 1865Q, Ozoace, Ozoace 0355, Ozoace 2701, P 116, P 127, PF 155, PX 100, Paracol 404A, Paracol 404C, Paracol 5000, Paracol 505N, Paradit P 60, Paradit PR, Paradit PR New, Paradit PR New A, Paradium SS, Paraffin 5203, Paraffin 6214, Parafilm, Paraflint C 80N6, Paraflint H 105, Paraflint H 4, Paraflint HIN 3, Paraflint Hi-Ni, Paraflint RG, Paraflint Spray 30, Paraflint Spray 40, Paralite 17, Paramelt 4608, Paranok 203, Paraseal, Paratex NS 70D, Parawax, Parvan 158, Parvan 161, Parvan 5250, Parvan Wax 6550, Permal, Petroleum waxes, Petrolite PX 100, Petrox P 200, Popko 5000, Popko 600, Press-Aid XP, Press-Aid-SP, Pt-White Wax 0602, Purunitto 50Z, R 152, R 152 (wax), R 4700, RCH Wax, RPW 1-85, RPW 1-89, RPW 1-94, RPW 2-92, RPW 2-93, RPW 2-94, Rheolube 185, SP 0110, SP 0140, SP 0145, SP 1004, SP 1030, SP 1030 (wax), SP 1044, SP 1044 (wax), SP 3035, SP 3040, SP 3040 (wax), SPW 116H, SR 143, SW 137, San Rogue Wax, Selosol 686, Shell 100, Shell 270, Shellmax, Shellwax 100, Shellwax 120, Shellwax 200, Shellwax 300, Shellwax 400, Sizepine W 116H, Slackwax 11, Slackwax 30, Slopvox, Smithwax 117, Sunnoc, Sunnoc B, Sunnoc N, Sunoco 4415, Sunoco 4417, Sunoco P 116, Sunwax 4412, Sunwax 5512, Swix CH 10, Swix CH 8, Syntene 160, Synwax 22XF, T 550, T 550 (wax), TH 44, Thermobank C, Topco S 904A, Ubatol EXP 21, Ultraflex, Ultraflex (wax), Ultralube W 7090, VOA 2080, Vebafine FT 300, Vebawax SH 105, Vestowax FT 200, Vestowax H 2, Vestowax SH 112, Vestowax SP 1024, Victory, Victory (wax), Vultex 8, Waxall A, Waxes and Waxy substances, hydrocarbon, Waxrex 2470, Witco 128, Witco 85010-1, Wosten 65, X 7905, XL 165, XL 165SB, YaV 1, ZV 1, ZV-P

Specifications

Quantity 500 g
Chemical Name or Material Paraffin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.