missing translation for 'onlineSavingsMsg'
Learn More

Paraffin Oils, TRC

Product Code. 30419718
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419718 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419718 Supplier Toronto Research Chemicals Supplier No. TRCP19328350ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mineral Oil
CAS 8012-95-1
Quantity 50 mL
Synonym Paraffin oil, 100 Solvent, 100S, 100S (paraffin), 150S, 260S, 350S, 40S, 40S (paraffin), 415 Oil, 55S, 60S, 60S (paraffin), 70S, 70S (paraffin), 7LT-NF, 80S, AES 150, AF 400, Abion E, Acme S 105, Actrel 3349L, Actrel 3360L, Actrel 4493L, Additol VXW 4932, Agrirob, Ampol C15 CPD, Ampol C23 DC-Tron NR, Arcoprime 200, Arcoprime 300, Arcoprime 350, Arcoprime 400, Arcoprime 70, Atplus 411F Oil, Atpolan, Atpolan 80EC, Avilub RS, BJ Oil, BP 620, BP Energol GR-XP 220, BP Energol WM 2, BVM 100N, BZ 1, Banole, Banole HV, Barrel Process Oil P 10000, Barrel Process Oil P 2200, Bayol 72, Bayol 82, Bayol 95, Behran 840, Bio-Dux, Biobase 300, Bonide, Brian R-N, Britol 20T, CEC-RL 175/3, COC, CP 14N, CP 15N, CP 40, CP 40 (oil), CP 50S, CP 68N, Caloria HT 43, Calpar 70, Cantenex SM 925, Capl 1, Capl 2, Carnation, Carnation 70, Catenex 946, Catenex 956, Catenex OR, Catenex P 491, Catenex P 941, Catenex S, Catenex S 932, Catenex SM 925, Catenex SNR, Catenex T, Catenex T 145, Certrex 60, Chevron 100R, Citation 70NY, Citgo 340 Neutral, Citrole, Clenvex AS 58, Coker gatch oil, Conosol C 200, Cosmo Neutral 500, Cosmo Pure Flex, Cosmo Purespin D, Cosmo Purespin G, Cosmo White P 70, Cosmol, Cosmol (oil), Crop oil, Crystal Plus 70T, Crystol 102, Crystol 172, Crystol 202, Crystol 322, Crystol 325, Crystol 352, Crystol 52, Crystol 542, Crystol 70, Crystol 72, Crystol J 142, Crystol J 262, Crystol J 352, Crystol J 52, Crystol J 72, Crystol N 352, Crystol N 72, Cypar 7, Cypar 9, D 110, D 130, D 60, D 70, D 80, D 95, DC 350, DE 69, DW 5052, DW 6266, DW 7580, DY 36, Dalian SN 70, Daphne 32N, Daphne AL 35, Daphne CP 12N, Daphne CP 15N, Daphne CP 50S, Daphne CP 68, Daphne CP 68N, Daphne FP 50, Daphne KP 100, Daphne KP 15, Daphne KP 32, Daphne KP 32U, Daphne KP 68, Daphne Oil CP, Daphne Oil CP 68N, Daphne Oil KP 100, Daphne Oil KP 32, Daphne Oil KP 68, Diana Fresia P 430, Diana Fresia PW 8, Diana Fresia PW 90, Diana Fresia S 32, Diana NU 80, Diana PM 90, Diana PS 32, Diana PS 380, Diana PS 430, Diana PS 90, Diana PW 100, Diana PW 32, Diana PW 380, Diana PW 90, Diana PW-LV, Diana PX 8, Diana Process Oil NR, Diana Process Oil P 430, Diana Process Oil PS 32, Diana Process Oil PS 380, Diana Process Oil PS 430, Diana Process Oil PS 90, Diana Process Oil PW 100, Diana Process Oil PW 32, Diana Process Oil PW 32AH24, Diana Process Oil PW 380, Diana Process Oil PW 90, Diana Solvent 2, Dicera 11833, Drakeol, Drakeol 10, Drakeol 10B, Drakeol 10LT, Drakeol 13, Drakeol 15, Drakeol 19, Drakeol 21, Drakeol 24, Drakeol 32, Drakeol 33, Drakeol 34, Drakeol 34NF, Drakeol 35, Drakeol 350, Drakeol 5, Drakeol 600, Drakeol 6VR, Drakeol 7, Drakeol 7LT-NF, Drakeol 7NF, Drakeol 9, Drakeol LT, Drakeol Supreme, Drakesol 260, Drakesol 260AT, Duoprime 200, Duoprime 350, Duoprime 400, Duoprime 500, Duoprime 70, Duoprime 90, E 340, E 340 (paraffin), E-C 100N, ED 320H, EDC 99DW, EM 80, EP-POIL, EZL 550, EZL 600, EZL 675, Edelex SM 925, Elevast C 30, Elevast C 70, Elevast E 10, Elevast E 20, Elevast R 120, Elevast R 130, Elevast R 150, Elevast R 170, Enerpar 13, Enerpar 1903, Enerpar M 1930, Enerpar M 2520, Escaid 90, Essobayol 90, Essotherm 500, Exchange, Extensoil 51-24T, Exxal D 110, Exxal D 60, Exxal D 80, Exxon 100N, Exxon D 60, Exxpar 252, Exxsol D 100, Exxsol D 100-140, Exxsol D 100S, Exxsol D 110, Exxsol D 130, Exxsol D 140, Exxsol D 160, Exxsol D 200, Exxsol D 220/230, Exxsol D 30, Exxsol D 40, Exxsol D 60, Exxsol D 70, Exxsol D 80, Exxsol D 95, Exxysn, F 9807, FBK Oil RO 100, FIH 6, Faxam 22, Fina F 9807, Finavestan A 50B, Finavestan A 80B, Fixaoil, Flavex 913, Flexon 815, Flexon 819, Flexon 845, Flexon 848, Flexon 865, Flexon 885, Fluid paraffins, Fractol A, Freezene 100, Freezene 200, Fukkol Process Oil P 100, Fukkol Process Oil P 200, Fukkol Process Oil P 400, G 400H, GQ-HVI Ib 500, Golden Bear Oil, Golden Bear Oil 1111, Gulf 150E, Gulf 2210, HB, HB (paraffin), HC 250, HC 250 (solvent), HC 370, HC-FX 50, HMVIP 30, HW 350, HW 350 (paraffin), Hi-Call K 230, Hi-Call K 350, Hi-Soflon, Paraffin oil, Acme S 105, Actrel 3349L, Actrel 3360L, Actrel 4493L, Additol VXW 4932, Agrirob, Ampol C15 CPD, Ampol C23 DC-Tron NR, Arcoprime 200, Atpolan, Atpolan 80EC, Avilub RS, BP Energol GR-XP 220, BP Energol WM 2, Banole, Banole HV, Barrel ProcessOil P 10000, Barrel Process Oil P 2200, Cactus Normal Paraffin YHNP, Caloria HT 43, Calpar 70, Cantenex SM 925, Carnation, Certrex 60, Chevron 100R, Citation 70NY, Citgo 340 Neutral, Citrole, Clenvex AS 58, Coker gatch oil, Conosol C 200, Cosmo Neutral 500, Crop oil, Crystal Plus 70T, Cypar 7, Dalian SN 70, Diana Fresia P 430, Dicera 11833, Drakeol, Duoprime, Edelex SM 925, Elevast, Enerpar, Escaid 90, Essobayol 90, Essotherm 500, Exchange, Exssol D 80, Extensoil 51-24T, Exxal, Exxsol, Exxysn, Finavestan A, Fixaoil, Flavex 913, Flexon, Fluid paraffins, Fractol A, Freezene, Fukkol Process Oil P 100, Hiclean CF 50, Hicol, Hydrorafinat H 5, Hydroseal G, Hydrowax, Hyper White 350, Hyprene P 150BS, Iberpesa T 601, Inland 19, Ipsol H, Irgawax, Isocer NL, JMS Stylet-Oil, JOMO Process, Kemesol, Kermac 500T, Kixx LP, Klearol, Klearol 50, Kolex 320, Komorex, Kornoil Concentrate, Kyowazol C 1200, Kyowazol C 900, Lacolene, Lacquer Diluent 6, Lamacit APM 520, Liquid Paraffin 60S, Lubinol, Lucant 360, MagieSol 44, Magnaflux, Manol, Marcol, Markol 82, Michang P 3, Miclube 1460, Mineral Seal Oil, Mineral oil','paraffinic, Mobilcer, Mobiltherm 603, Moresco P 40, Nisseki Hi-White 70, Nopco 1407, Normal Paraffin TS 6, Normal paraffin, NovaSpec 2050, Nujol, O-WO2, Octafoam, Octopus P Plus 3029, Oleo FC, Ondina 15, Organic JMS Stylet-Oil, Osco Baby Oil, Pale Oil 60, Parabase, Paracos KF 350, Paraffin, Paraflex, Paralux, Paramil MES 15, Parax 40K3, Parex II, Parol 70, Parprol 22, Parprol 32, Peneteck, PennzUltra, Peptoil, Perleam 18, Petrolatum oil, Petrolatum','liq., Pharma 240B/HV6TI, Phazol 35, Pioneer 2002, Pionier 2024, Plastol 355, Plastol 537, Plaxene 185, Primol, Process oils
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.