missing translation for 'onlineSavingsMsg'
Learn More

Paraffin Liquid, Pure, Water White, d=0.83-0.86, Fisher Chemical™

Product Code. 10589190 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic container
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10589190 25 L Plastic container 25 L
10655692 2.5 L Amber glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10589190 Supplier Fisher Chemical Supplier No. P/0400/25
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Paraffin Liquid, Pure, Water White, d=0.83-0.86

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number 131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point -9°C
Boiling Point 310°C
Packaging Plastic container
Quantity 25 L
Vapor Density >2 at 101kPa
Viscosity >19 St (40°C)
Physical Form Viscous Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.