missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Paraffin Dissolver

Product Code. 11942422
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 11942422

Brand: Macherey-Nagel™ 740968.25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) For the Removal of Paraffin from FFPE Sections
Product Type Paraffin Dissolver
Quantity 25 mL
compliance-icons
Product Identifier
  • Paraffin Dissolver (25 mL)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : paraffin disolver
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.