missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Paraffin Dissolver (blue)

Product Code. 15809077
Change view
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15809077 60 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809077 Supplier Macherey-Nagel™ Supplier No. 740343.60

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paraffin Dissolver to be used with NucleoSpin totalRNA FFPE, NucleoSpin totalRNA FFPE XS, NucleoSpin 96 DNA FFPE, or NucleoMag DNA FFPE kits

NucleoSpin™ 8/96 DNA FFPE Kit is used for isolation of total DNA from formalin-fixed, paraffin-embedded (FFPE) samples using either a flexible 8-well strip format for varying throughput or a proven 96-well plate format for high troughput.

  • Rapid isolation of DNA from fresh and archived FFPE samples, specimen of object slides
  • Blue colored Paraffin Dissolver (patented technology) for easy paraffin removal
  • Decrosslinking buffer to eliminate formalin crosslinking
  • High quality DNA for improved performance in downstream applications (e.g., PCR)
  • Processing under vacuum or by centrifugation, suitable for manual and automated processing

Specifications

CAS 64742-46-7
Color Blue
For Use With (Application) Paraffin removal during DNA isolation of FFPE samples
For Use With (Equipment) NucleoSpin 8/96 DNA FFPE kit
Physical Form Liquid
Quantity 60 mL
Formulation Paraffin dissolver, Blue dye
Odor Organic
Solubility Information <0.001% in Water
compliance-icons
Product Identifier
  • Paraffin Dissolver (blue) (60 mL)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: paraffin disolver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.