missing translation for 'onlineSavingsMsg'
Learn More

Paracetamol, Lipomed

Product Code. 30461389
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461389 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461389 Supplier Lipomed Supplier No. LPMPAR28450
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Paracetamol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paracetamol
CAS 103-90-2
Analyte or Component Names Paracetamol
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Synonym Acetamide, N-(4-hydroxyphenyl)-, Acetanilide, 4'-hydroxy-, 4'-Hydroxyacetanilide, 4-(Acetylamino)phenol, 4-(N-Acetylamino)phenol, 4-Acetamidophenol, 4-Acetaminophenol, 4-Hydroxyacetanilide, APAP, Abensanil, Acamol, Acenol, Acenol (pharmaceutical), Acetagesic, Acetalgin, Acetaminofen, Acetaminophen, Algotropyl, Alpiny, Alvedon, Amadil, Anaflon, Anelix, Anhiba, Apamid, Apamide, Banesin, Ben-u-ron, Bickie-mol, Biocetamol, Calpol, Captin, Cetadol, Citramon P, Claratal, Clixodyne, Crocin, Dafalgan, Daga, Daphalgan, Datril, Dial-a-gesic, Dirox, Disprol, Doliprane, Dolprone, Duorol, Dymadon, Efferalgan, Endophy, Enelfa, Eneril, Eu-Med, Exdol, Febrilex, Febrilix, Febro-Gesic, Febrolin, Fendon, Fepanil, Finimal, Gattaphen T, Gelocatil, Hedex, Homoolan, Jin Gang, Korum, Lestemp, Liquagesic, Lonarid, Lyteca, Lyteca Syrup, Momentum, Multin, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, N-Acetyl-4-hydroxyaniline, N-Acetyl-p-aminophenol, NAPA, NAPA (analgesic), NAPAP, NSC 109028, NSC 3991, Napafen, Naprinol, Nebs, Nobedon, Ortensan, Pacemo, Pacemol, Paldesic, Pamol, Panadol, Panadol Actifast, Panadol Extend, Panaleve, Panasorb, Panets, Panodil, Paracetamol, Paracetamol DC, Paracetamole, Paralen, Paramol, Paraspen, Parelan, Parmol, Pasolind N, Pedric, Perfalgan, Phenaphen, Phendon, Pinex, Pinex (pharmaceutical), Prodafalgan, Puerxitong, Pyrinazine, Resfenol, Resprin, Rhodapop NCR, Salzone, Sara, Tabalgin, Tachipirina, Tapar, Temlo, Tempanal, Tempra, Tralgon, Tylenol, Valadol, Valgesic, Vick Pyrena, p-(Acetylamino)phenol, p-Aceaminophenol, p-Acetamidophenol, p-Acetoaminophen, p-Hydroxyacetanilide, panadeine
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(=O)Nc1ccc(O)cc1
IUPAC Name N-(4-hydroxyphenyl)acetamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 151.16
Quantity 50 mg
Formula Weight 151.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.