missing translation for 'onlineSavingsMsg'
Learn More

Paracetamol EP Impurity J (Acetaminophen USP Related Compound J), Mikromol™

Product Code. 17990916
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990916 25 mg 25mg
17904336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17990916 Supplier Mikromol Supplier No. MM0042.020025
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Paracetamol EP Impurity J (Acetaminophen USP Related Compound J)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloroacetanilide
CAS 539-03-7
Analyte or Component Names 4-Chloroacetanilide
Shipping Condition Room Temperature
Molecular Formula C8 H8 Cl N O
Certifications/Compliance ISO 17025
Synonym N-(4-Chlorophenyl)acetamide (4-Chloroacetanilide), N-(4-Chlorophenyl)acetamide, Paracetamol Imp. J (EP), 4-Chloroacetanilide, Acetaminophen USP Related Compound J, Acetaminophen USP RC J, SKF-8835, USP Acetaminophen Related Compound J, Ph Eur Paracetamol Impurity J, Acetaminophen Related Compound J, Paracetamol Impurity J
InChI Formula InChI=1S/C8H8ClNO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11)
SMILES CC(=O)Nc1ccc(Cl)cc1
IUPAC Name N-(4-chlorophenyl)acetamide
Molecular Weight (g/mol) 169.61
Quantity 25 mg
Formula Weight 169.0294 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.