missing translation for 'onlineSavingsMsg'
Learn More

Paraben target analyte mix solution, Fluka™

Product Code. 16242892 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 16242892

Brand: Honeywell-Fluka R11475ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

10 μg/mL in acetone, analytical standard, reference material

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paraben target analyte mix solution
Packaging Glass Bottle
CAS 1416711-53-9, 67-64-1, 1581694-95-2
Color Colorless
Melting Point -95°C (Data refer to the solvent.)
Boiling Point 55°C to 57°C (1.013 hPa) (Data refer to the solvent.)
Quantity 5 mL
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.