missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAR6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15847613
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847613 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15847613 Supplier Invitrogen™ Supplier No. PA569202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 87%; Yeast: 85%.

This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 domain and a semi-Cdc42/Rac interactive binding domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PARD6A
Gene Accession No. Q9Z101
Gene Alias 0710008C04Rik; 2610010A15Rik; PAR6; Par-6; par-6 (partitioning defective 6,) homolog alpha; PAR-6 alpha; par-6 family cell polarity regulator alpha; par-6 partitioning defective 6 homolog alpha; PAR6A; PAR-6A; PAR6alpha; PAR6C; Pard6a; Partitioning defective 6 homolog alpha; partitioning defective protein 6A; partitioning defective-6 homolog alpha; partitioning-defective protein 6; similar to Partitioning defective-6 homolog alpha (PAR-6 alpha) (PAR-6A) (PAR-6) (PAR6C) (Tax interaction protein 40) (TIP-40); tax interaction protein 40; TAX40; Tax-interacting protein 40; Tip-40
Gene Symbols PARD6A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse PAR6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56513
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.