missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAR6 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 domain and a semi-Cdc42/Rac interactive binding domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Antigen | PAR6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PARD6A |
| Gene Accession No. | Q9NPB6, Q9Z101 |
| Gene Alias | 0710008C04Rik; 2610010A15Rik; PAR6; Par-6; par-6 (partitioning defective 6,) homolog alpha; PAR-6 alpha; par-6 family cell polarity regulator alpha; par-6 partitioning defective 6 homolog alpha; PAR6A; PAR-6A; PAR6alpha; PAR6C; Pard6a; Partitioning defective 6 homolog alpha; partitioning defective protein 6A; partitioning defective-6 homolog alpha; partitioning-defective protein 6; similar to Partitioning defective-6 homolog alpha (PAR-6 alpha) (PAR-6A) (PAR-6) (PAR6C) (Tax interaction protein 40) (TIP-40); tax interaction protein 40; TAX40; Tax-interacting protein 40; Tip-40 |
| Gene Symbols | PARD6A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?