missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PAR4 Monoclonal antibody specifically detects PAR4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
Specifications
| Antigen | PAR4 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 5F4 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_003941 |
| Gene Alias | proteinase-activated receptor 4, coagulation factor II (thrombin) receptor-like 3, PAR4 |
| Host Species | Mouse |
| Immunogen | F2RL3 (NP_003941, 18 a.a. ~ 82 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GGTQTPSVYDESGSTGGGDDSTPSILPAPRGYPGQVCANDSDTLELPDSSRALLLGWVPTRLVPA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?