missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PAR2 Antibody (Regeneron patent anti-PAR-2), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28886-50ug
This item is not returnable.
View return policy
Description
PAR2 Monoclonal antibody specifically detects PAR2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PAR2 | |
| Monoclonal | |
| Unconjugated | |
| P55085 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Regeneron patent anti-PAR-2 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1 | |
| PAR2 | |
| 50 μg | |
| GPCR, Signal Transduction | |
| 2150 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction