Learn More
Novus Biologicals™ PAR2 Antibody (PAR650097), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28757-100ug
 View more versions of this product
Product Code. 30044553
  • £266.00 / 100µg
Description
PAR2 Monoclonal antibody specifically detects PAR2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAR2
Monoclonal
Unconjugated
P55085
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
PAR650097
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1
PAR2
100 μg
GPCR, Signal Transduction
2150
Human
Purified