missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PAR2 Antibody Blocking Peptide

Product Code. 18696461 Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
This item is not returnable. View return policy

Product Code. 18696461

Brand: Novus Biologicals™ NBP211250PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene ID (Entrez) 2150
Species Mouse
Purity >85%, by HPLC
Concentration 1.2 mg/mL
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP2-11250
Gene Alias coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1
Gene Symbol F2RL1
Quantity 0.25 mg
Research Category GPCR, Signal Transduction
Storage Buffer Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.