missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR1/Thrombin Receptor Antibody (N2-11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£174.00 - £433.00
Specifications
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100. Use reported in scientific literature (PMID 35055169), Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31885791), Immunofluorescence |
|---|---|
| Classification | Monoclonal |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Molecular Weight of Antigen | 52 kDa |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18717023
|
Novus Biologicals
NBP1-71770SS |
£174.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18728283
|
Novus Biologicals
NBP1-71770 |
£433.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100. Use reported in scientific literature (PMID 35055169), Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31885791), Immunofluorescence | |
| Monoclonal | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title