missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR1/Thrombin Receptor Antibody (5E2E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£163.00 - £435.00
Specifications
| Antigen | PAR1/Thrombin Receptor |
|---|---|
| Clone | 5E2E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253534
|
Novus Biologicals
NBP2-61770 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604418
|
Novus Biologicals
NBP2-61770-0.025ml |
0.025 mL |
£163.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PAR1/Thrombin Receptor | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1 | |
| F2R | |
| IgG1 | |
| Protein G purified |
| 5E2E6 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Cancer, GPCR | |
| 2149 | |
| Purified recombinant fragment of human F2R (AA: 42-176) expressed in E. Coli. | |
| Primary | |
| 47.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title