missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR/pADPr Antibody (10H), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-89039
This item is not returnable.
View return policy
Description
PAR/pADPr Monoclonal specifically detects PAR/pADPr in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PAR/pADPr | |
| Monoclonal | |
| Unconjugated | |
| 50mM HEPES, pH 7.4, containing 100mM sodium chloride, 1% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human, Mouse, Rat, Drosophila | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 10H | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-20 ug/ml, Immunohistochemistry-Paraffin 5-20 ug/ml | |
| Poly(ADP-ribose) | |
| Purified poly(ADP-ribose). | |
| 50 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction