missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAQR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569886
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
PAQR4 gene ontology annotations for this gene involves signaling receptor activity.
Specifications
| PAQR4 | |
| Polyclonal | |
| Unconjugated | |
| PAQR4 | |
| 1500004C10Rik; LOW QUALITY PROTEIN: progestin and adipoQ receptor family member 4; MNCb-0914; Paqr4; Progestin and adipoQ receptor family member 4; progestin and adipoQ receptor family member IV | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 302967 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| PAQR4 | |
| synthetic peptide directed towards the middle region of human PAQR4. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction