missing translation for 'onlineSavingsMsg'
Learn More

PAPSS2 Antibody (OTI6H10) [PE], Novus Biologicals™

Product Code. 30687878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687878 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687878 Supplier Novus Biologicals Supplier No. NBP344692PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PAPSS2 Monoclonal antibody specifically detects PAPSS2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPSS2
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI6H10
Conjugate PE
Formulation PBS
Gene Alias 3'-phosphoadenosine 5'-phosphosulfate synthase 2, ATP sulfurylase/adenosine 5'-phosphosulfate kinase, ATPSK2ATP sulfurylase/APS kinase 2, bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2, bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2, EC 2.7.1.25, PAPS synthase 2, PAPS synthetase 2, PAPSS 2, SK 2, SK2, Sulfurylase kinase 2,3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-247 of human PAPSS2(NP_004661) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9060
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.