missing translation for 'onlineSavingsMsg'
Learn More

PAPSS2 Antibody, Novus Biologicals™

Product Code. 18379488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379488 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379488 Supplier Novus Biologicals Supplier No. H00009060B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PAPSS2 Polyclonal antibody specifically detects PAPSS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPSS2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.4)
Gene Accession No. NP_004661.2
Gene Alias 3'-phosphoadenosine 5'-phosphosulfate synthase 2, ATP sulfurylase/adenosine 5'-phosphosulfate kinase, ATPSK2ATP sulfurylase/APS kinase 2, bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2, bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2, EC 2.7.1.25, PAPS synthase 2, PAPS synthetase 2, PAPSS 2, SK 2, SK2, Sulfurylase kinase 2,3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2
Host Species Mouse
Immunogen PAPSS2 (NP_004661.2, 1 a.a. - 614 a.a.) full-length human protein. MSGIKKQKTENQQKSTNVVYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTISFALEEYLVSHAIPCYSLDGDNVRHGLNRNLGFSPGDREENIRRIAEVAKLFADAGLVCITSFISPFAKDRENARKIHESAGLPFFEIFVDAPLNICESRDVKGLYKRARAGEIKGFTGIDSDYEKPETPERVLKTNLSTVSDCVHQVVELLQEQNIVPYTIIKDIHELFVPENKLDHVRAEAETLPSLSITKLDLQWVQVLSEGWATPLKGFMREKEYLQVMHFDTLLDDGVINMSIPIVLPVSAEDKTRLEGCSKFVLAHGGRRVAILRDAEFYEHRKEERCSRVWGTTCTKHPHIKMVMESGDWLVGGDLQVLEKIRWNDGLDQYRLTPLELKQKCKEMNADAVFAFQLRNPVHNGHALLMQDTRRRLLERGYKHPVLLLHPLGGWTKDDDVPLDWRMKQHAAVLEEGVLDPKSTIVAIFPSPMLYAGPTEVQWHCRSRMIAGANFYIVGRDPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLEKN
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9060
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.