missing translation for 'onlineSavingsMsg'
Learn More

PAPSS1 Antibody (1F4), Novus Biologicals™

Product Code. 18327518 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327518 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327518 Supplier Novus Biologicals Supplier No. H00009061M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PAPSS1 Monoclonal antibody specifically detects PAPSS1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPSS1
Applications Western Blot, ELISA, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 1F4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005434
Gene Alias 3'-phosphoadenosine 5'-phosphosulfate synthase 1, ATPSK1SK 1, bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 1, EC 2.7.1.25, PAPS synthase 1, PAPSS 1, PAPSS3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 1, SK1, Sulfurylase kinase 1
Host Species Mouse
Immunogen PAPSS1 (NP_005434, 144 a.a. ~ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 9061
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.