Learn More
GVS PAPR High Efficiency (HE) Filter
Product Code. 18409625
Change view
Click to view available options
Cartridge Type:
P3
Unit Size:
Each
Product Code. Cartridge Type unitSize
18409625 P3 Each
1 options
Product Code. 18409625 Supplier GVS Supplier No. 03892P
The PX5™ P3 particulate filters protect the user in a range of environments from industrial to medical.
The PX5™ P3 Particulate filters protect the user in a range of environments from industrial to medical.
  • Lightweight, compact and easy to replace.
  • Can filter 99.97% of particles down to 0.03 of a micron
  • Certified to EN 12941 1998 +A2:2008
  • Includes: PX5™ P3 Particulate Filter
Specifications
Cartridge Type P3
Color Gray
Material Plastic
Product Type Cartridge
Size One Size