missing translation for 'onlineSavingsMsg'
Learn More

Pappalysin-2/PAPP-A2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18414717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414717 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414717 Supplier Novus Biologicals Supplier No. NBP176487AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pappalysin-2/PAPP-A2 Polyclonal antibody specifically detects Pappalysin-2/PAPP-A2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pappalysin-2/PAPP-A2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24.-, PAPP-A2pappalysin-2, pappalysin 2, PAPPE, PAPP-E, PLAC3, placenta-specific 3, pregnancy-associated plasma preproprotein-A2, Pregnancy-associated plasma protein A2, Pregnancy-associated plasma protein E1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human PLAC3. The immunogen is located within amino acids 450 - 500 of PLAC3. Amino Acid Squence: VPFRDEKYPRLEVLQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 60676
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.