missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (9), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
£429.00
Specifications
| Antigen | Pappalysin-1/PAPP-A |
|---|---|
| Clone | 9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | PerCP |
Description
Pappalysin-1/PAPP-A Monoclonal antibody specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Pappalysin-1/PAPP-A | |
| Western Blot | |
| PerCP | |
| Mouse | |
| Cancer | |
| PBS | |
| 5069 | |
| IgG1 κ | |
| Protein A or G purified |
| 9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
| Purified human PAPP-A | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title