missing translation for 'onlineSavingsMsg'
Learn More

Pappalysin-1/PAPP-A Antibody (9), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18414715 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414715 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414715 Supplier Novus Biologicals Supplier No. NBP105116MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pappalysin-1/PAPP-A Monoclonal antibody specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pappalysin-1/PAPP-A
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 9
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1
Host Species Mouse
Immunogen Purified human PAPP-A
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5069
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.