missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAPPA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553798
This item is not returnable.
View return policy
Description
Immunogen sequence: GDSSEDGHYF RGHLGTLVFW STALPQSHFQ HSSQHSSGEE EATDLVLTAS FEPVNTEWVP FRDEKYPRLE VLQGFE Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 64%.
PLAC3 (placenta-specific protein 3), also known as PAPP-A2, is a metalloproteinase that cleaves the insulin-like growth factor-binding protein (IGFBP)-5 in a IGF-independent fashion. It has been suggested that the cleavage of IGFBP-5 and to a lesser extent IGFBP-3 produced in the maternal decidua promotes feto-placental growth. Recent studies have shown elevated levels of PLAC3 in hypertensive disorders of pregnancy including preeclampsia and HELLP (hemolysis, elevated liver enzymes, and low platelets) syndrome.
Specifications
| PAPPA2 | |
| Polyclonal | |
| Unconjugated | |
| PAPPA2 | |
| LOC680415; PAPPA2; PAPP-A2; pappalysin 2; pappalysin-2; Pappe; PAPP-E; PLAC3; placenta-specific 3; pregnancy-associated plasma preproprotein-A2; Pregnancy-associated plasma protein A2; Pregnancy-associated plasma protein E1; pregnancy-associated plasma protein-E; RP4-774I24.1; similar to pappalysin 2 isoform 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60676 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXP8 | |
| PAPPA2 | |
| Recombinant protein corresponding to Human PAPPA2. Recombinant protein control fragment (Product #RP-91868). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction