missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPPA, Mouse anti-Human, Clone: 1G3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005069-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: GATEEPSPPSRALYFSGRGEQLRLRADLELPRDAFTLQVWLRAEGGQRSPAVITGLYDKCSYISRDRGWVVGIHTISDQDNKDPRYFFSLKTDRARQVTTINAHRSYLPGSpecifications
| PAPPA | |
| Monoclonal | |
| Unconjugated | |
| pregnancy-associated plasma protein A, pappalysin 1 | |
| ASBABP2/DIPLA1/IGFBP-4ase/PAPA/PAPP-A/PAPPA1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1G3 | |
| In 1x PBS, pH 7.4 | |
| NM_002581 | |
| PAPPA | |
| PAPPA (NP_002572, 85 a.a. ∼ 194 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5069 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction