missing translation for 'onlineSavingsMsg'
Learn More

PAPOLG Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18414923 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414923 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414923 Supplier Novus Biologicals Supplier No. NBP130060JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PAPOLG Polyclonal antibody specifically detects PAPOLG in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPOLG
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.7.19, FLJ11805, FLJ12972, FLJ13482, FLJ14187, MGC133307, MGC133308, neo-PAP, Neo-poly(A) polymerase, nuclear poly(A) polymerase gamma, PAP2, PAPG, PAP-gamma, poly(A) polymerase gamma, Polynucleotide adenylyltransferase gamma, SRP RNA 3' adenylating enzyme/pap2, SRP RNA 3'-adenylating enzyme
Host Species Rabbit
Immunogen maps to a region between residue 625 and 675 of human poly(A) polymerase gamma using the numbering given in entry NP_075045.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64895
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.