Learn More
Invitrogen™ PAPOLB Monoclonal Antibody (3D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522298
Description
Peptide Sequence: MMPFPVTTQG PPQPAPPPNR YGVSSPISLA VPKETDCLLT QRLIETLRPF GVFEEEEELQ RRILVLDKLN NLVKEWIREI SESKSLPQSV IENVGGKIFT FGSYRLGVHT KGADIDALCV APSHVDRSDF FTSFYAKLKL QEEVKDLRAV QEAFVPVIKL CFDGIEIDIL FARLALQTIP EDLDLRDDSL LKNLDIRCIR SLNGCRVTDE ILHLVPNIDN FRLTLRAIKL WAKCHNIYSN ILGFLGGVSW AMLVARTCQL YPNAVASTLV RKFFLVFSEW EWPNPVLLKE PEERNLNLPV WDPRVNPSDR YHLMPIITPA YPQQNSTYNV SISTRMVMIE EFKQGLAITH EILLSKAEWS KLFEAPSFFQ KYKHYIVLLA SASTEKQHLE WVGLVESKIR ILVGSLEKNE FITLAHVNPQ SFPAPKENPD MEEFRTMWVI GLGLKKPDNS EILSIDLTYD IQSFTDTVYR QAVNSKMFEM GMKITAVHLR RKELHQLLPH HVLQDKKAHS TEGRRLTDLN DSSFDLSAGC ENSMSVPSST STMKTGPLIS SSQGRNSPAL AVMTASVANI QATEFSLQQV NTNESSGVAL NESIPHAVSQ PAISPSPKAM VARVVSSTCL ISHPDLQETQ QQTYLIL.
Testis-specific poly(A) polymerase.
Specifications
| PAPOLB | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NRJ5 | |
| PAPOLB | |
| PAPOLB (AAH36653, 1 a.a. ~ 637 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3D7 | |
| Unconjugated | |
| PAPOLB | |
| Med21; PAP; PAP-beta; Paplob; Papola-ps; PAPOLB; PAPT; Plap-ps; poly (A) polymerase beta (testis specific); poly(A) polymerase beta; poly(A) polymerase beta (testis specific); polynucleotide adenylyltransferase beta; Testis-specific poly(A) polymerase; TPAP | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56903 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.