missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPOLA Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
PAPOLA Polyclonal antibody specifically detects PAPOLA in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | PAPOLA |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:500 |
| Formulation | Whole antiserum |
| Gene Accession No. | P51003 |
| Gene Alias | EC 2.7.7, EC 2.7.7.19, PAP-alpha, PAPMGC5378, poly(A) polymerase alpha, Polynucleotide adenylyltransferase alpha |
| Host Species | Rabbit |
| Immunogen | Polyclonal antibody raised in rabbit against human PAPOLA (polyA polymerase alpha), using a KLH-conjugated synthetic peptide containing a sequence from the C-terminal part of the protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?